Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DHX16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164239.1 | NP_001157711.1 | 5 | 646 | ||
NM_003587.4 | NP_003578.2 | 5 | 967 | |||
XM_011514938.2 | XP_011513240.1 | 2 | 760 | |||
XM_011514939.2 | XP_011513241.1 | 2 | 234 | |||
XM_011514940.2 | XP_011513242.1 | 4 | 499 | |||
GenBank mRNA | AB001601.1 | - | 5 | 894 | ||
AK302721.1 | - | 5 | 640 | |||
AK312264.1 | - | 5 | 906 | |||
BC008825.2 | - | 5 | 918 | 4100320 | ||
BC009392.2 | - | 5 | 918 | 4136069 |