Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CUL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003592.2 | NP_003583.2 | 15, 16 | 2025 | ||
XM_005250060.4 | XP_005250117.1 | 15, 16 | 2061 | |||
XM_011516629.2 | XP_011514931.1 | 15, 16 | 2017 | |||
XM_011516630.2 | XP_011514932.1 | 15, 16 | 1918 | |||
XM_011516631.2 | XP_011514933.1 | 15, 16 | 2046 | |||
XM_011516632.2 | XP_011514934.1 | 15, 16 | 1944 | |||
XM_017012723.1 | XP_016868212.1 | 14, 15 | 1781 | |||
GenBank mRNA | AF062536.1 | - | 14, 15 | 1799 | ||
AK096163.1 | - | 16, 17 | 1913 | |||
AK096505.1 | - | 8, 9 | 1682 | |||
AK296909.1 | - | 14, 15 | 1750 | |||
AK315023.1 | - | 15, 16 | 2201 | |||
BC125119.1 | - | 15, 16 | 1938 | 40118923 | ||
BC125120.1 | - | 15, 16 | 1938 | 40118927 | ||
BX537409.1 | - | 15, 16 | 1955 | |||
U58087.1 | - | 15, 16 | 1727 |