Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATRN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001207047.2 | NP_001193976.1 | 8 | 1028 | ||
NM_001323332.1 | NP_001310261.1 | 8 | 1411 | |||
NM_139321.2 | NP_647537.1 | 8 | 1411 | |||
NM_139322.3 | NP_647538.1 | 8 | 1411 | |||
GenBank mRNA | AF106861.1 | - | 8 | 1321 | ||
AK293010.1 | - | 8 | 1411 | |||
AK302730.1 | - | 8 | 1007 | |||
BC101705.1 | - | 8 | 1361 | 8069211 |