Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPST2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008566.1 | NP_001008566.1 | 3 | 1094 | ||
NM_003595.3 | NP_003586.3 | 3 | 986 | |||
XM_006724338.3 | XP_006724401.1 | 4 | 1105 | |||
GenBank mRNA | AF049891.1 | - | 3 | 950 | ||
AF061254.1 | - | 1 | 758 | |||
AJ006198.1 | - | 1 | 840 | |||
AK074538.1 | - | 3 | 986 | |||
AK075139.1 | - | 3 | 1058 | |||
AK304545.1 | - | 5 | 1070 | |||
AL136623.2 | - | 3 | 970 | |||
BC001057.2 | - | 3 | 965 | 3509627 | ||
BC017509.1 | - | 3 | 950 | 4857366 | ||
CR456597.1 | - | 1 | 848 | |||
CR533528.1 | - | 1 | 753 |