Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPM1D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003620.3 | NP_003611.1 | 2, 3 | 936 | ||
GenBank mRNA | AK299773.1 | - | 3, 4 | 645 | ||
AK312902.1 | - | 2, 3 | 926 | |||
BC016480.1 | - | 2, 3 | 852 | 3923593 | ||
BC033893.2 | - | 2, 3 | 810 | 5265521 | ||
BC042418.1 | - | 2, 3 | 850 | 5167004 | ||
BC060877.1 | - | 2, 3 | 936 | 5260081 | ||
BT009780.1 | - | 2, 3 | 704 | |||
KU178420.1 | - | 2, 3 | 704 | |||
U78305.1 | - | 2, 3 | 907 |