Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITGA10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303040.1 | NP_001289969.1 | 17 | 2331 | ||
NM_001303041.1 | NP_001289970.1 | 16 | 2219 | |||
NM_003637.4 | NP_003628.2 | 20 | 2648 | |||
XM_005277436.3 | XP_005277493.1 | 19 | 2399 | |||
XM_011510083.2 | XP_011508385.1 | 16 | 2079 | |||
XM_011510084.1 | XP_011508386.1 | 9 | 1076 | |||
XM_017002622.1 | XP_016858111.1 | 18 | 2605 | |||
XM_017002623.1 | XP_016858112.1 | 18 | 2602 | |||
XM_017002624.1 | XP_016858113.1 | 18 | 2605 | |||
XM_017002625.1 | XP_016858114.1 | 18 | 2605 | |||
XM_017002626.1 | XP_016858115.1 | 16 | 2362 | |||
XM_017002627.1 | XP_016858116.1 | 18 | 2672 | |||
XM_017002628.1 | XP_016858117.1 | 14 | 2055 | |||
GenBank mRNA | AF074015.1 | - | 20 | 2494 | ||
AF112345.1 | - | 20 | 2549 | |||
AK304155.1 | - | 17 | 2187 | |||
AK304183.1 | - | 19 | 2478 | |||
AK314255.1 | - | 20 | 2497 | |||
BC140831.1 | - | 16 | 2094 | 9021699 | ||
BC144637.1 | - | 16 | 2094 | 9053168 |