Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITGA10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303040.1 | NP_001289969.1 | 24 | 3041 | ||
NM_001303041.1 | NP_001289970.1 | 23 | 2929 | |||
NM_003637.4 | NP_003628.2 | 27 | 3358 | |||
XM_005277436.3 | XP_005277493.1 | 26 | 3109 | |||
XM_011510083.2 | XP_011508385.1 | 23 | 2789 | |||
XM_011510084.1 | XP_011508386.1 | 16 | 1786 | |||
XM_017002622.1 | XP_016858111.1 | 25 | 3315 | |||
XM_017002623.1 | XP_016858112.1 | 25 | 3312 | |||
XM_017002625.1 | XP_016858114.1 | 24 | 3201 | |||
XM_017002626.1 | XP_016858115.1 | 23 | 3072 | |||
XM_017002628.1 | XP_016858117.1 | 21 | 2765 | |||
GenBank mRNA | AF074015.1 | - | 27 | 3204 | ||
AF112345.1 | - | 27 | 3259 | |||
AK304155.1 | - | 24 | 2897 | |||
AK314255.1 | - | 27 | 3207 | |||
BC140831.1 | - | 23 | 2804 | 9021699 | ||
BC144637.1 | - | 23 | 2804 | 9053168 |