Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CBX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003655.2 | NP_003646.2 | 5 | 613 | ||
XM_011525399.2 | XP_011523701.1 | 3 | 2122 | |||
GenBank mRNA | AF013956.1 | - | 5 | 610 | ||
BG029608.1 | - | 5 | 445 | |||
EU439707.1 | - | 5 | 645 | |||
KU178423.1 | - | 5 | 450 |