Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLF7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270942.1 | NP_001257871.1 | 2 | 298 | ||
NM_003709.3 | NP_003700.1 | 1 | 508 | |||
NR_073108.1 | 2 | 472 | ||||
XM_005246926.3 | XP_005246983.1 | 2 | 294 | |||
XM_006712815.3 | XP_006712878.1 | 2 | 298 | |||
XM_011512072.2 | XP_011510374.1 | 2 | 472 | |||
XM_011512073.2 | XP_011510375.1 | 2 | 261 | |||
XM_011512075.1 | XP_011510377.1 | 1 | 481 | |||
XM_017005161.1 | XP_016860650.1 | 2 | 294 | |||
XM_017005163.1 | XP_016860652.1 | 1 | 481 | |||
XM_017005164.1 | XP_016860653.1 | 1 | 493 | |||
GenBank mRNA | AB015132.1 | - | 1 | 383 | ||
AK027371.1 | - | 1 | 209 | |||
AK314439.1 | - | 1 | 439 | |||
AL042506.1 | - | 1 | 510 | |||
BC012919.1 | - | 2 | 289 | 4509200 | ||
BP229510.1 | - | 1 | 510 | |||
CR542112.1 | - | 1 | 60 | |||
DA023826.1 | - | 2 | 268 | |||
DA800775.1 | - | 2 | 298 | |||
HF546205.1 | - | 1 | 261 | |||
HF546206.1 | - | 1 | 261 |