Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSD17B6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003725.3 | NP_003716.2 | 5 | 1030 | ||
XM_005269207.1 | XP_005269264.1 | 6 | 1179 | |||
XM_005269208.1 | XP_005269265.1 | 6 | 1183 | |||
XM_005269209.1 | XP_005269266.1 | 6 | 1170 | |||
XM_006719672.1 | XP_006719735.1 | 6 | 1178 | |||
XM_011538925.1 | XP_011537227.1 | 6 | 1173 | |||
XM_011538926.1 | XP_011537228.1 | 6 | 1163 | |||
XM_011538927.1 | XP_011537229.1 | 6 | 1154 | |||
GenBank mRNA | AF016509.1 | - | 4 | 814 | ||
AF223225.1 | - | 5 | 901 | |||
AI760144.1 | - | 1 | 592 | |||
AK313423.1 | - | 5 | 940 | |||
BC020710.1 | - | 5 | 924 | 4700726 | ||
U89281.1 | - | 5 | 919 |