Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAH11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277115.1 | NP_001264044.1 | 63 | 10340 | ||
GenBank mRNA | AJ320497.1 | - | 63 | 10361 |