Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B4GALT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199873.1 | NP_001186802.1 | 7 | 1604 | ||
NM_001199874.1 | NP_001186803.1 | 7 | 1533 | |||
NM_003779.3 | NP_003770.1 | 7 | 1583 | |||
XM_005245566.1 | XP_005245623.1 | 8 | 1305 | |||
XM_011510093.1 | XP_011508395.1 | 7 | 1139 | |||
XM_017002714.1 | XP_016858203.1 | 7 | 1160 | |||
GenBank mRNA | AB024435.1 | - | 5 | 882 | ||
AF038661.1 | - | 7 | 1110 | |||
AK023311.1 | - | 8 | 1068 | |||
AK056851.1 | - | 5 | 2271 | |||
AK092493.1 | - | 8 | 1065 | |||
BC000276.1 | - | 7 | 1086 | 3357342 | ||
BC006099.1 | - | 7 | 1100 | 3677118 | ||
BC009985.2 | - | 7 | 1062 | 4124756 | ||
Y12509.1 | - | 5 | 877 |