Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GBF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199378.1 | NP_001186307.1 | 22 | 3065 | ||
NM_001199379.1 | NP_001186308.1 | 22 | 3062 | |||
NM_004193.2 | NP_004184.1 | 22 | 3062 | |||
XM_005270261.1 | XP_005270318.1 | 22 | 3065 | |||
XM_006718047.1 | XP_006718110.1 | 23 | 3164 | |||
XM_006718048.1 | XP_006718111.1 | 23 | 3161 | |||
XM_006718049.1 | XP_006718112.1 | 23 | 3164 | |||
XM_006718050.1 | XP_006718113.1 | 23 | 3128 | |||
XM_011540312.1 | XP_011538614.1 | 22 | 3023 | |||
XM_011540313.1 | XP_011538615.1 | 23 | 3164 | |||
XM_011540314.2 | XP_011538616.1 | 11 | 1542 | |||
XM_017016861.1 | XP_016872350.1 | 23 | 3140 | |||
XM_017016862.1 | XP_016872351.1 | 23 | 3128 | |||
XM_017016863.1 | XP_016872352.1 | 22 | 3026 | |||
XM_017016864.1 | XP_016872353.1 | 18 | 2364 | |||
GenBank mRNA | AF068755.1 | - | 22 | 3008 | ||
AK024916.1 | - | 2 | 175 | |||
AK025330.1 | - | 22 | 3031 | |||
BC117681.1 | - | 22 | 2889 | 40070852 | ||
BC117682.1 | - | 22 | 2889 | 40070853 | ||
D87435.2 | - | 22 | 2992 |