Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYOM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003803.3 | NP_003794.3 | 5 | 1166 | ||
NM_019856.1 | NP_062830.1 | 5 | 1166 | |||
XM_017026062.1 | XP_016881551.1 | 5 | 929 | |||
GenBank mRNA | AJ621424.1 | - | 5 | 1166 | ||
AK125322.1 | - | 5 | 1166 | |||
AK302444.1 | - | 3 | 507 | |||
BC116183.1 | - | 5 | 907 | 40073728 | ||
X69090.1 | - | 3 | 541 |