Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PEX11B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001184795.1 | NP_001171724.1 | 3 | 352 | ||
NM_003846.2 | NP_003837.1 | 3 | 546 | |||
NR_073491.1 | 3 | 542 | ||||
NR_073492.1 | 3 | 536 | ||||
NR_073493.2 | 3 | 758 | ||||
GenBank mRNA | AB018080.1 | - | 3 | 328 | ||
AF093670.1 | - | 3 | 326 | |||
AK023991.1 | - | 3 | 310 | |||
AK301983.1 | - | 3 | 312 | |||
BC011963.1 | - | 3 | 326 | 3866690 | ||
BG423552.1 | - | 286 | ||||
CB155824.1 | - | 3 | 352 | |||
CR542047.1 | - | 3 | 310 | |||
DB054933.1 | - | 3 | 546 | |||
HY108328.1 | - | 3 | 329 |