Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: INPP4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001101669.1 | NP_001095139.1 | 15 | 1655 | ||
NM_003866.2 | NP_003857.2 | 16 | 1754 | |||
XM_005263323.3 | XP_005263380.1 | 8 | 863 | |||
XM_005263325.3 | XP_005263382.1 | 8 | 836 | |||
XM_011532391.2 | XP_011530693.1 | 16 | 1766 | |||
XM_011532392.2 | XP_011530694.1 | 15 | 1505 | |||
XM_017008794.1 | XP_016864283.1 | 16 | 1766 | |||
XM_017008795.1 | XP_016864284.1 | 16 | 1766 | |||
XM_017008796.1 | XP_016864285.1 | 15 | 1739 | |||
XM_017008797.1 | XP_016864286.1 | 9 | 886 | |||
XM_017008798.1 | XP_016864287.1 | 9 | 863 | |||
GenBank mRNA | AK298118.1 | - | 8 | 804 | ||
AK300891.1 | - | 8 | 869 | |||
AK302936.1 | - | 14 | 1399 | |||
BC072447.1 | - | 14 | 1562 | |||
BC110918.1 | - | 14 | 1512 | 6160633 | ||
BC133005.1 | - | 12 | 1210 | 40147027 | ||
BC144255.1 | - | 12 | 1210 | |||
BX649090.1 | - | 16 | 1595 | |||
U96922.1 | - | 12 | 1280 |