Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ARHGEF7 SIRNA Assays ›
Gene Symbol:
ARHGEF7
Gene Name:
Rho guanine nucleotide exchange factor 7
Gene Aliases:
BETA-PIX, COOL-1, COOL1, Nbla10314, P50, P50BP, P85, P85COOL1, P85SPR, PAK3, PIXB
Chromosome Location:
Chr.13: 111114619 - 111305734 on Build GRCh38
UniGene ID:
Hs.508738
Species:
Human
siRNA ID
HSS113109
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ARHGEF7
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001113511.2
NP_001106983.1
6
1504
NM_001113512.2
NP_001106984.1
4
1354
NM_001113513.1
NP_001106985.1
4
446
NM_001320851.1
NP_001307780.1
3
227
NM_001320852.1
NP_001307781.1
5
1441
NM_001320853.1
NP_001307782.1
4
352
NM_003899.3
NP_003890.1
4
559
NM_145735.3
NP_663788.1
5
1441
XM_006719956.3
XP_006720019.1
5
569
XM_011521133.2
XP_011519435.1
5
569
XM_017020814.1
XP_016876303.1
5
569
XM_017020815.1
XP_016876304.1
3
562
XM_017020816.1
XP_016876305.1
4
337
XM_017020817.1
XP_016876306.1
3
562
XM_017020818.1
XP_016876307.1
5
437
XM_017020819.1
XP_016876308.1
4
446
XM_017020820.1
XP_016876309.1
4
569
XM_017020821.1
XP_016876310.1
4
264
XM_017020822.1
XP_016876311.1
3
260
XM_017020823.1
XP_016876312.1
3
1037
XM_017020824.1
XP_016876313.1
5
437
XM_017020825.1
XP_016876314.1
3
188
XM_017020826.1
XP_016876315.1
5
437
XM_017020827.1
XP_016876316.1
3
261
GenBank mRNA
AB177848.1
-
5
1442
AB177849.1
-
2
165
AK300157.1
-
4
270
AK300272.1
-
4
339
AL834228.1
-
5
625
BC033905.1
-
4
551
5262912
BC050521.1
-
4
418
BC060776.1
-
6
846
5271977
D63476.1
-
4
535
DA216264.1
-
4
446
DA404477.1
-
5
437
DA439385.1
-
6
524
DC318118.1
-
4
352
Back To Top
More Information
Panther Classification:
Molecular Function -
guanyl-nucleotide exchange factor
Gene Ontology Categories:
Function(s)
Process(es)
hematopoietic progenitor cell differentiation
Golgi organization
signal transduction
nervous system development
positive regulation of fibroblast migration
lamellipodium assembly
positive regulation of protein binding
regulation of Rho protein signal transduction
intracellular signal transduction
negative regulation of epidermal growth factor receptor signaling pathway
positive regulation of apoptotic process
positive regulation of GTPase activity
ephrin receptor signaling pathway
focal adhesion assembly
regulation of small GTPase mediated signal transduction
positive regulation of growth hormone secretion
positive regulation of substrate adhesion-dependent cell spreading
regulation of GTP binding
positive regulation of lamellipodium morphogenesis
guanyl-nucleotide exchange factor activity
Rho guanyl-nucleotide exchange factor activity
protein binding
protein kinase binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-567f868b4f-c74kb:80/10.239.167.51:80.
git-commit: 96b0dd45ad55c22ec1de1c352dff25df4a817ab2
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.8.0-2024.05.19-1.0