Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GYG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079855.1 | NP_001073324.1 | 11 | 1575 | ||
NM_001184702.1 | NP_001171631.1 | 11 | 1572 | |||
NM_001184703.1 | NP_001171632.1 | 10 | 1455 | |||
NM_001184704.1 | NP_001171633.1 | 8 | 1220 | |||
NM_003918.2 | NP_003909.2 | 12 | 1668 | |||
XM_011545600.2 | XP_011543902.1 | 11 | 1506 | |||
XM_017029927.1 | XP_016885416.1 | 12 | 1618 | |||
XM_017029928.1 | XP_016885417.1 | 12 | 1593 | |||
XM_017029929.1 | XP_016885418.1 | 12 | 1775 | |||
XM_017029930.1 | XP_016885419.1 | 9 | 1270 | |||
XM_017029931.1 | XP_016885420.1 | 9 | 1293 | |||
GenBank mRNA | AK098046.1 | - | 8 | 1128 | ||
AK292496.1 | - | 11 | 1483 | |||
BC023152.1 | - | 11 | 1442 | 3923041 | ||
DQ424901.1 | - | 8 | 1173 | |||
U94357.1 | - | 2 | 122 | |||
U94360.1 | - | 6 | 910 | |||
U94362.1 | - | 12 | 1669 | |||
U94363.1 | - | 11 | 1446 |