Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HERC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271602.1 | NP_001258531.1 | 19 | 2275 | ||
NM_014606.2 | NP_055421.1 | 20 | 2530 | |||
XM_005263327.3 | XP_005263384.1 | 21 | 2657 | |||
XM_005263329.3 | XP_005263386.1 | 19 | 2506 | |||
XM_005263330.3 | XP_005263387.1 | 19 | 2359 | |||
XM_005263331.1 | XP_005263388.1 | 20 | 2530 | |||
XM_005263332.3 | XP_005263389.1 | 20 | 2530 | |||
XM_011532397.2 | XP_011530699.1 | 7 | 770 | |||
XM_017008806.1 | XP_016864295.1 | 20 | 2633 | |||
XM_017008807.1 | XP_016864296.1 | 18 | 2229 | |||
XM_017008808.1 | XP_016864297.1 | 13 | 1432 | |||
GenBank mRNA | AK289990.1 | - | 20 | 2473 | ||
AK296379.1 | - | 19 | 2251 | |||
AK296397.1 | - | 8 | 994 | |||
D25215.1 | - | 20 | 2457 |