Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SELENBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001258288.1 | NP_001245217.1 | 11 | 1310 | ||
NM_001258289.1 | NP_001245218.1 | 12 | 1705 | |||
NM_003944.3 | NP_003935.2 | 12 | 1496 | |||
GenBank mRNA | AK023875.1 | - | 8 | 2587 | ||
AK296661.1 | - | 12 | 1674 | |||
AK298355.1 | - | 12 | 1361 | |||
AK303815.1 | - | 11 | 1276 | |||
AK315643.1 | - | 12 | 1466 | |||
BC009084.1 | - | 12 | 1450 | 3853674 | ||
BC032997.1 | - | 10 | 1881 | 4839079 | ||
BQ045424.1 | - | 1 | 269 | |||
CR456852.1 | - | 11 | 1403 | |||
EU794660.1 | - | 12 | 1450 | |||
U29091.1 | - | 11 | 1407 |