Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAAR5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003967.2 | NP_003958.2 | 1 | 989 | ||
GenBank mRNA | AF021818.1 | - | 1 | 937 | ||
BC069171.1 | - | 1 | 937 | 7216924 | ||
BC095541.1 | - | 1 | 937 | 30706570 | ||
BC112209.1 | - | 1 | 989 | 8327677 | ||
BC112211.1 | - | 1 | 989 | 8327679 |
Set Membership: |
G-protein coupled receptor |