Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267580.1 | NP_001254509.1 | 11 | 1614 | ||
NM_003981.3 | NP_003972.1 | 12 | 1737 | |||
NM_199413.2 | NP_955445.1 | 12 | 1737 | |||
XM_005254987.2 | XP_005255044.1 | 12 | 1645 | |||
XM_006720759.1 | XP_006720822.1 | 12 | 1645 | |||
XM_006720760.1 | XP_006720823.1 | 12 | 1645 | |||
XM_011522187.1 | XP_011520489.1 | 12 | 1645 | |||
XM_011522188.2 | XP_011520490.1 | 12 | 1645 | |||
XM_011522190.2 | XP_011520492.1 | 11 | 1474 | |||
XM_011522191.2 | XP_011520493.1 | 12 | 1645 | |||
XM_011522192.1 | XP_011520494.1 | 10 | 1324 | |||
XM_017022714.1 | XP_016878203.1 | 11 | 1519 | |||
XM_017022715.1 | XP_016878204.1 | 11 | 1519 | |||
XM_017022716.1 | XP_016878205.1 | 11 | 1474 | |||
XM_017022717.1 | XP_016878206.1 | 11 | 1522 | |||
GenBank mRNA | AF044588.1 | - | 12 | 1581 | ||
AK297117.1 | - | 11 | 1488 | |||
AK312600.1 | - | 12 | 1635 | |||
BC003138.1 | - | 12 | 1605 | 2958690 | ||
BC005140.2 | - | 12 | 1545 | 3504761 | ||
BG823292.1 | - | 2 | 89 | |||
BX647317.1 | - | 3 | 273 |