Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM50A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004699.3 | NP_004690.1 | 9, 10 | 895 | ||
GenBank mRNA | AD001530.1 | - | 9, 10 | 860 | ||
AK293119.1 | - | 9, 10 | 849 | |||
AK313697.1 | - | 9, 10 | 843 | |||
BC000028.2 | - | 9, 10 | 866 | 3505365 | ||
BU679052.1 | - | 4, 5 | 471 | |||
D83260.1 | - | 9, 10 | 765 |