Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EBAG9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278938.1 | NP_001265867.1 | 6 | 564 | ||
NM_004215.4 | NP_004206.1 | 6 | 851 | |||
NM_198120.2 | NP_936056.1 | 6 | 684 | |||
XM_017013960.1 | XP_016869449.1 | 6 | 1047 | |||
GenBank mRNA | AB007619.1 | - | 6 | 732 | ||
AF006265.1 | - | 6 | 682 | |||
AK290651.1 | - | 6 | 673 | |||
AL515533.3 | - | 6 | 742 | |||
AY515724.1 | - | 6 | 642 | |||
AY653072.1 | - | 5 | 449 | |||
BC005249.1 | - | 6 | 762 | 3930583 | ||
BC017729.1 | - | 6 | 635 | 3453385 | ||
BC022506.1 | - | 6 | 749 | 4815654 | ||
BG616704.1 | - | 6 | 565 |