Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZBED1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001171135.1 | NP_001164606.1 | 2 | 509 | ||
NM_001171136.1 | NP_001164607.1 | 2 | 431 | |||
NM_004729.3 | NP_004720.1 | 2 | 428 | |||
GenBank mRNA | AB018328.1 | - | 2 | 428 | ||
BC015030.1 | - | 2 | 509 | 3908412 | ||
Y16947.1 | - | 1 | 283 |