Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other DLGAP1 SIRNA Assays ›
Gene Symbol:
DLGAP1
Gene Name:
DLG associated protein 1
Gene Aliases:
DAP-1, DAP-1-ALPHA, DAP-1-BETA, DAP1, DLGAP1A, DLGAP1B, GKAP, SAPAP1
Chromosome Location:
Chr.18: 3496032 - 4455307 on Build GRCh38
UniGene ID:
Hs.594640
Species:
Human
siRNA ID
HSS113679
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: DLGAP1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001003809.2
NP_001003809.1
3
406
NM_001242761.1
NP_001229690.1
6
1763
NM_001242762.1
NP_001229691.1
3
433
NM_001242763.1
NP_001229692.1
3
433
NM_001242764.1
NP_001229693.1
3
379
NM_001242765.1
NP_001229694.1
3
406
NM_001242766.1
NP_001229695.1
3
406
NM_001308390.1
NP_001295319.1
3
431
NM_004746.3
NP_004737.2
6
1763
XM_005258171.2
XP_005258228.1
6
1804
XM_005258172.1
XP_005258229.1
6
1799
XM_005258173.3
XP_005258230.2
4
595
XM_005258174.4
XP_005258231.1
3
652
XM_006722367.3
XP_006722430.1
4
763
XM_011525770.2
XP_011524072.1
6
1804
XM_011525771.2
XP_011524073.1
6
1804
XM_017026080.1
XP_016881569.1
4
762
XM_017026081.1
XP_016881570.1
3
426
XM_017026082.1
XP_016881571.1
4
581
XM_017026083.1
XP_016881572.1
3
651
XM_017026084.1
XP_016881573.1
3
648
XM_017026085.1
XP_016881574.1
3
641
GenBank mRNA
AB000276.1
-
3
349
AB000277.1
-
3
1303
AK294717.1
-
3
433
AK294747.1
-
3
406
AK294777.1
-
3
356
AK299880.1
-
3
379
AK304101.1
-
6
1745
AK308827.1
-
3
382
AK316099.1
-
6
1763
BC070175.1
-
3
1244
6620806
BC136453.1
-
3
1295
9020442
BC136454.1
-
3
1295
9020443
U67988.1
-
3
353
Back To Top
More Information
Panther Classification:
Molecular Function -
scaffold/adaptor protein
Gene Ontology Categories:
Function(s)
Process(es)
chemical synaptic transmission
protein binding
protein domain specific binding
protein complex binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-86c4h:80/10.239.166.160:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0