Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB11B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004218.3 | NP_004209.2 | 4 | 565 | ||
GenBank mRNA | AF498947.1 | - | 4 | 469 | ||
AK225036.1 | - | 4 | 483 | |||
AK225642.1 | - | 4 | 485 | |||
AK297498.1 | - | 4 | 493 | |||
AK310573.1 | - | 4 | 256 | |||
AK312994.1 | - | 4 | 519 | |||
BC110081.1 | - | 4 | 471 | 40035112 | ||
BT019535.1 | - | 4 | 469 | |||
BT019536.1 | - | 4 | 469 | |||
BX647356.1 | - | 2567 | ||||
CR541691.1 | - | 4 | 469 | |||
CX787570.1 | - | 2 | 116 | |||
D38516.1 | - | 4 | 478 | |||
EF560724.1 | - | 4 | 548 | |||
X79780.1 | - | 4 | 475 |