Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TRIP12 SIRNA Assays ›
Gene Symbol:
TRIP12
Gene Name:
thyroid hormone receptor interactor 12
Gene Aliases:
TRIP-12, ULF
Chromosome Location:
Chr.2: 229763837 - 229923234 on Build GRCh38
UniGene ID:
Hs.572642
Species:
Human
siRNA ID
HSS113807
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TRIP12
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001284214.1
NP_001271143.1
21
3186
NM_001284215.1
NP_001271144.1
19
3011
NM_001284216.1
NP_001271145.1
18
2232
NM_004238.2
NP_004229.1
20
3042
XM_005246954.4
XP_005247011.1
21
3294
XM_005246955.4
XP_005247012.1
21
3293
XM_005246956.4
XP_005247013.1
21
3270
XM_005246957.4
XP_005247014.1
21
3260
XM_005246958.4
XP_005247015.1
21
3213
XM_005246960.4
XP_005247017.1
22
3207
XM_005246961.3
XP_005247018.1
20
3144
XM_005246962.4
XP_005247019.1
20
3141
XM_006712852.3
XP_006712915.1
20
3097
XM_011512180.2
XP_011510482.1
21
3247
XM_011512181.2
XP_011510483.1
21
3245
XM_011512183.2
XP_011510485.1
20
3101
XM_017005272.1
XP_016860761.1
22
3353
XM_017005273.1
XP_016860762.1
21
3220
XM_017005274.1
XP_016860763.1
21
3168
XM_017005275.1
XP_016860764.1
21
3223
XM_017005276.1
XP_016860765.1
21
3228
XM_017005277.1
XP_016860766.1
22
3368
XM_017005278.1
XP_016860767.1
21
3183
XM_017005279.1
XP_016860768.1
21
3255
XM_017005280.1
XP_016860769.1
21
3244
XM_017005281.1
XP_016860770.1
21
3163
XM_017005282.1
XP_016860771.1
22
3206
XM_017005283.1
XP_016860772.1
21
3210
XM_017005284.1
XP_016860773.1
22
3204
XM_017005285.1
XP_016860774.1
20
3131
XM_017005286.1
XP_016860775.1
20
3163
XM_017005287.1
XP_016860776.1
20
3144
XM_017005288.1
XP_016860777.1
20
3106
XM_017005289.1
XP_016860778.1
22
3126
XM_017005290.1
XP_016860779.1
22
3123
XM_017005291.1
XP_016860780.1
21
3057
XM_017005292.1
XP_016860781.1
20
3060
XM_017005293.1
XP_016860782.1
20
3060
GenBank mRNA
BC113891.1
-
18
2126
40083155
BC114556.1
-
21
3080
40083165
D28476.1
-
20
2972
EU489742.1
-
19
2965
Back To Top
More Information
Panther Classification:
Molecular Function -
ubiquitin-protein ligase
Gene Ontology Categories:
Function(s)
Process(es)
protein polyubiquitination
DNA repair
cellular response to DNA damage stimulus
embryo development
protein ubiquitination involved in ubiquitin-dependent protein catabolic process
negative regulation of histone H2A K63-linked ubiquitination
negative regulation of double-strand break repair
ubiquitin-protein transferase activity
protein binding
ligase activity
thyroid hormone receptor binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wjc8w:80/10.239.165.44:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0