Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CER1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005454.2 | NP_005445.1 | 2 | 696 | ||
GenBank mRNA | AF400435.1 | - | 2 | 651 | ||
BC069371.1 | - | 2 | 651 | |||
BC069491.1 | - | 2 | 651 | 7262148 | ||
BC069503.1 | - | 2 | 651 | 7262160 | ||
BC103976.1 | - | 2 | 651 | 40015380 | ||
BC103977.1 | - | 2 | 651 | 40015381 | ||
BU071333.1 | - | 1 | 519 |