Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADIPOQ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177800.1 | NP_001171271.1 | 4 | 796 | ||
NM_004797.3 | NP_004788.1 | 3 | 745 | |||
GenBank mRNA | AK291525.1 | - | 3 | 718 | ||
AK312868.1 | - | 4 | 766 | |||
AX767959.1 | - | 3 | 687 | |||
BC054496.1 | - | 686 | 6192794 | |||
BC096308.3 | - | 3 | 729 | 40007540 | ||
BC096309.3 | - | 3 | 729 | 40007541 | ||
BC096310.3 | - | 3 | 729 | 40007542 | ||
BC096311.3 | - | 3 | 729 | 40007544 | ||
D45371.1 | - | 3 | 687 | |||
EU420013.1 | - | 2 | 661 |
Gene Symbol: ADIPOQ-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046662.1 | 1 | 1490 | |||
GenBank mRNA | BC036509.1 | - | 2 | 1626 |