Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MAP4K4 SIRNA Assays ›
Gene Symbol:
MAP4K4
Gene Name:
mitogen-activated protein kinase kinase kinase kinase 4
Gene Aliases:
FLH21957, HEL-S-31, HGK, MEKKK4, NIK
Chromosome Location:
Chr.2: 101697703 - 101894690 on Build GRCh38
UniGene ID:
Hs.701013
Species:
Human
siRNA ID
HSS114054
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MAP4K4
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001242559.1
NP_001229488.1
27
3225
NM_001242560.1
NP_001229489.1
28
3586
NM_004834.4
NP_004825.3
27
3352
NM_145686.3
NP_663719.2
28
3700
NM_145687.3
NP_663720.1
28
3144
XM_005264048.2
XP_005264105.1
30
3729
XM_005264050.3
XP_005264107.1
29
3567
XM_005264052.3
XP_005264109.1
29
3564
XM_005264057.3
XP_005264114.1
29
3498
XM_005264058.3
XP_005264115.1
28
3489
XM_005264059.3
XP_005264116.1
29
3405
XM_005264060.3
XP_005264117.1
28
3393
XM_005264062.3
XP_005264119.1
28
3336
XM_005264063.3
XP_005264120.1
28
3333
XM_005264064.3
XP_005264121.1
27
3327
XM_005264065.3
XP_005264122.1
28
3243
XM_005264066.3
XP_005264123.1
28
3240
XM_005264068.3
XP_005264125.1
27
3234
XM_005264069.2
XP_005264126.1
27
3204
XM_005264071.2
XP_005264128.1
26
3132
XM_006712865.2
XP_006712928.1
29
3474
XM_006712868.2
XP_006712931.1
28
3243
XM_006712869.2
XP_006712932.1
27
3144
XM_017005349.1
XP_016860838.1
30
3633
XM_017005350.1
XP_016860839.1
29
3567
XM_017005351.1
XP_016860840.1
28
3486
XM_017005352.1
XP_016860841.1
29
3498
XM_017005353.1
XP_016860842.1
29
3495
XM_017005354.1
XP_016860843.1
29
3474
XM_017005355.1
XP_016860844.1
28
3465
XM_017005356.1
XP_016860845.1
28
3396
XM_017005357.1
XP_016860846.1
29
3405
XM_017005358.1
XP_016860847.1
29
3402
XM_017005359.1
XP_016860848.1
28
3375
XM_017005360.1
XP_016860849.1
28
3372
XM_017005361.1
XP_016860850.1
28
3375
XM_017005362.1
XP_016860851.1
28
3372
XM_017005363.1
XP_016860852.1
28
3336
XM_017005364.1
XP_016860853.1
27
3324
XM_017005365.1
XP_016860854.1
28
3306
XM_017005366.1
XP_016860855.1
28
3282
XM_017005367.1
XP_016860856.1
28
3279
XM_017005368.1
XP_016860857.1
28
3210
XM_017005369.1
XP_016860858.1
27
3141
XM_017005370.1
XP_016860859.1
27
3141
XM_017005371.1
XP_016860860.1
26
3135
XM_017005372.1
XP_016860861.1
27
3051
XM_017005373.1
XP_016860862.1
26
3042
XM_017005374.1
XP_016860863.1
27
3051
XM_017005375.1
XP_016860864.1
26
3042
GenBank mRNA
AB013385.1
-
11
1132
AB014587.1
-
26
3028
AF096300.1
-
27
3053
AK001272.1
-
11
1287
AK074592.1
-
7
893
AK295610.1
-
27
3370
AK296396.1
-
28
3282
AL137755.1
-
20
2681
AY212247.1
-
27
3220
AY834276.1
-
23
2629
EU794634.1
-
28
3194
Back To Top
More Information
Panther Classification:
Molecular Function -
non-receptor serine/threonine protein kinase
Gene Ontology Categories:
Function(s)
Process(es)
protein phosphorylation
signal transduction by protein phosphorylation
microvillus assembly
intracellular signal transduction
negative regulation of apoptotic process
regulation of JNK cascade
creatine kinase activity
protein serine/threonine kinase activity
receptor signaling protein serine/threonine kinase activity
protein binding
ATP binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-fwm9x:80/10.239.164.221:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0