Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LY86 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004271.3 | NP_004262.1 | 3 | 296 | ||
XM_017011505.1 | XP_016866994.1 | 3 | 358 | |||
GenBank mRNA | AB020499.1 | - | 2 | 132 | ||
AF057178.2 | - | 3 | 285 | |||
AY931019.1 | - | 3 | 262 | |||
BC038846.1 | - | 3 | 306 | 6015123 | ||
CD695164.1 | - | 3 | 360 |