Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SH3BP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001018009.3 | NP_001018009.2 | 5 | 693 | ||
NM_004844.4 | NP_004835.2 | 5 | 812 | |||
XM_011534251.2 | XP_011532553.1 | 5 | 624 | |||
XM_011534253.1 | XP_011532555.1 | 5 | 773 | |||
XM_017007521.1 | XP_016863010.1 | 5 | 607 | |||
XM_017007522.1 | XP_016863011.1 | 6 | 1250 | |||
XM_017007523.1 | XP_016863012.1 | 5 | 1028 | |||
XM_017007524.1 | XP_016863013.1 | 5 | 560 | |||
XM_017007525.1 | XP_016863014.1 | 5 | 474 | |||
GenBank mRNA | AB005047.1 | - | 5 | 564 | ||
AK090524.1 | - | 7 | 916 | |||
AK313167.1 | - | 5 | 770 | |||
AL117422.1 | - | 5 | 768 | |||
AW673562.1 | - | 3 | 377 | |||
BC006169.1 | - | 5 | 667 | |||
BC010123.2 | - | 5 | 743 | 3841593 | ||
BC023195.1 | - | 5 | 484 | |||
BG475051.1 | - | 632 |