Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCYT1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163265.1 | NP_001156737.1 | 1 | 294 | ||
NM_004845.4 | NP_004836.2 | 1 | 294 | |||
GenBank mRNA | AF052510.1 | - | 1 | 121 | ||
AF148464.1 | - | 1 | 65 | |||
AK315323.1 | - | 1 | 294 | |||
BG942820.1 | - | 1 | 193 |