Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AKAP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004274.4 | NP_004265.3 | 13 | 6041 | ||
XM_011537378.2 | XP_011535680.1 | 8 | 3505 | |||
XM_011537379.2 | XP_011535681.1 | 7 | 3442 | |||
XM_011537383.2 | XP_011535685.1 | 5 | 2893 | |||
XM_017021808.1 | XP_016877297.1 | 12 | 5908 | |||
XM_017021809.1 | XP_016877298.1 | 12 | 7281 | |||
XM_017021810.1 | XP_016877299.1 | 13 | 7471 | |||
XM_017021811.1 | XP_016877300.1 | 7 | 9182 | |||
GenBank mRNA | AB002309.2 | - | 13 | 5981 | ||
BC137232.1 | - | 13 | 5924 | 9021229 | ||
BC137233.1 | - | 13 | 5924 | 9021230 | ||
BC150288.1 | - | 13 | 5981 | 8860126 | ||
U17195.2 | - | 13 | 5997 |