Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SNCAIP SIRNA Assays ›
Gene Symbol:
SNCAIP
Gene Name:
synuclein alpha interacting protein
Gene Aliases:
SYPH1, Sph1
Chromosome Location:
Chr.5: 122311354 - 122464219 on Build GRCh38
UniGene ID:
Hs.426463
Species:
Human
siRNA ID
HSS114373
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SNCAIP
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001242935.2
NP_001229864.1
5
521
NM_001308100.1
NP_001295029.1
9
3018
NM_001308105.1
NP_001295034.1
5
1177
NM_001308106.1
NP_001295035.1
5
446
NM_001308107.1
NP_001295036.1
5
521
NM_001308108.1
NP_001295037.1
5
485
NM_001308109.1
NP_001295038.1
4
407
NM_005460.3
NP_005451.2
7
1573
NR_131761.1
7
1498
NR_131762.1
6
452
XM_005272138.3
XP_005272195.1
7
1496
XM_005272139.1
XP_005272196.1
7
1436
XM_006714734.2
XP_006714797.1
7
1629
XM_011543737.2
XP_011542039.1
8
1631
XM_011543738.2
XP_011542040.1
8
1630
XM_011543739.1
XP_011542041.1
8
1556
XM_011543741.2
XP_011542043.1
8
1580
XM_011543743.2
XP_011542045.1
8
2005
XM_011543745.1
XP_011542047.1
7
1460
XM_011543749.2
XP_011542051.1
5
811
XM_011543750.1
XP_011542052.1
4
289
XM_017010078.1
XP_016865567.1
8
3958
XM_017010079.1
XP_016865568.1
8
1631
XM_017010080.1
XP_016865569.1
8
1631
XM_017010081.1
XP_016865570.1
8
1629
XM_017010082.1
XP_016865571.1
7
1460
XM_017010083.1
XP_016865572.1
5
437
XM_017010084.1
XP_016865573.1
8
1637
XM_017010085.1
XP_016865574.1
6
516
XM_017010086.1
XP_016865575.1
8
1635
GenBank mRNA
AB110788.1
-
6
1311
AB110789.1
-
5
1131
AB110790.1
-
7
1452
AB110791.1
-
5
400
AB110792.1
-
4
259
AB110793.1
-
5
439
AB110794.1
-
3
145
AF076929.1
-
7
1404
AK001617.1
-
3
257
AK298882.1
-
5
402
AK299687.1
-
6
452
AK304646.1
-
4
239
AK310835.1
-
7
1405
BC033743.1
-
5
341
5193089
BC040552.1
-
9
2880
5298588
BC094759.1
-
5
399
4838145
BM908442.1
-
465
DQ227317.1
-
5
465
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
cell death
dopamine metabolic process
cellular protein metabolic process
regulation of neurotransmitter secretion
regulation of inclusion body assembly
protein binding
ubiquitin protein ligase binding
identical protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wzvj9:80/10.239.161.253:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0