Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TOX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014729.2 | NP_055544.1 | 4 | 780 | ||
XM_017014085.1 | XP_016869574.1 | 3 | 799 | |||
GenBank mRNA | AB018351.1 | - | 4 | 782 | ||
BC016665.1 | - | 4 | 623 | 4342384 |