Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZFYVE16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105251.2 | NP_001098721.1 | 16 | 4373 | ||
NM_001284236.1 | NP_001271165.1 | 16 | 4299 | |||
NM_014733.4 | NP_055548.3 | 15 | 4245 | |||
XM_005248632.4 | XP_005248689.1 | 15 | 4371 | |||
XM_011543753.2 | XP_011542055.1 | 16 | 4270 | |||
XM_017010091.1 | XP_016865580.1 | 17 | 4431 | |||
XM_017010092.1 | XP_016865581.1 | 16 | 4417 | |||
XM_017010093.1 | XP_016865582.1 | 15 | 2751 | |||
XM_017010094.1 | XP_016865583.1 | 14 | 2696 | |||
XM_017010095.1 | XP_016865584.1 | 17 | 4325 | |||
XM_017010096.1 | XP_016865585.1 | 12 | 1773 | |||
GenBank mRNA | AB002303.1 | - | 16 | 4313 | ||
AF434817.1 | - | 14 | 4065 | |||
AK127003.1 | - | 11 | 1462 | |||
AL833087.2 | - | 13 | 3292 | |||
BX537424.1 | - | 15 | 4197 | |||
BX649172.1 | - | 4 | 2847 | |||
CR933621.1 | - | 16 | 4247 |