Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIEZO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142864.3 | NP_001136336.2 | 14 | 2053 | ||
GenBank mRNA | AB161230.1 | - | 3 | 504 | ||
BC150271.1 | - | 3 | 344 | 9007243 | ||
D87071.1 | - | 3 | 344 | |||
KC602455.1 | - | 14 | 1797 |