Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001207033.1 | NP_001193962.1 | 5 | 1447 | ||
NM_001207034.1 | NP_001193963.1 | 5 | 1447 | |||
NM_001286572.2 | NP_001273501.1 | 5 | 808 | |||
NM_014771.3 | NP_055586.1 | 5 | 1447 | |||
XM_011545997.1 | XP_011544299.1 | 5 | 788 | |||
GenBank mRNA | AB014561.1 | - | 5 | 719 | ||
AF122819.1 | - | 5 | 723 | |||
AK027406.1 | - | 5 | 737 | |||
AK075526.1 | - | 3 | 468 | |||
AK291666.1 | - | 5 | 737 | |||
BC006133.1 | - | 5 | 714 | 3628183 | ||
BC011769.2 | - | 5 | 714 | 3607432 | ||
BC018647.2 | - | 5 | 714 | 2967275 | ||
BX640763.1 | - | 2 | 81 | |||
CR627431.1 | - | 5 | 776 | |||
KU178492.1 | - | 4 | 627 | |||
KU178493.1 | - | 4 | 627 |