Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIAA0141 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142603.2 | NP_001136075.1 | 8, 9 | 1017 | ||
NM_014773.4 | NP_055588.3 | 8, 9 | 1017 | |||
XM_005268547.3 | XP_005268604.1 | 8, 9 | 1052 | |||
XM_005268548.3 | XP_005268605.1 | 8, 9 | 1052 | |||
XM_005268552.2 | XP_005268609.1 | 8, 9 | 1052 | |||
XM_006714812.3 | XP_006714875.1 | 2, 3 | 358 | |||
XM_011537715.1 | XP_011536017.1 | 8, 9 | 1052 | |||
GenBank mRNA | AK292869.1 | - | 8, 9 | 966 | ||
BC007855.2 | - | 8, 9 | 993 | 4298364 | ||
BC010452.1 | - | 8, 9 | 908 | 4155782 | ||
BC011269.1 | - | 8, 9 | 908 | 4155755 | ||
BC011926.1 | - | 8, 9 | 938 | 4543650 | ||
BC035486.1 | - | 8, 9 | 933 | |||
D50931.1 | - | 8, 9 | 975 |