Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GIT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135213.1 | NP_001128685.1 | 4 | 543 | ||
NM_001135214.1 | NP_001128686.1 | 4 | 543 | |||
NM_014776.3 | NP_055591.2 | 4 | 543 | |||
NM_057169.3 | NP_476510.1 | 4 | 543 | |||
NM_057170.3 | NP_476511.1 | 4 | 543 | |||
NM_139201.2 | NP_631940.2 | 4 | 543 | |||
XM_005253997.3 | XP_005254054.1 | 4 | 538 | |||
XM_005254000.3 | XP_005254057.1 | 4 | 538 | |||
XM_006719707.3 | XP_006719770.1 | 4 | 538 | |||
XM_006719708.3 | XP_006719771.1 | 4 | 538 | |||
XM_006719709.3 | XP_006719772.1 | 4 | 538 | |||
XM_006719712.3 | XP_006719775.1 | 4 | 538 | |||
XM_006719713.3 | XP_006719776.1 | 4 | 535 | |||
XM_006719714.3 | XP_006719777.1 | 4 | 538 | |||
XM_017020258.1 | XP_016875747.1 | 4 | 538 | |||
XM_017020259.1 | XP_016875748.1 | 4 | 535 | |||
XM_017020260.1 | XP_016875749.1 | 4 | 535 | |||
XM_017020261.1 | XP_016875750.1 | 4 | 533 | |||
GenBank mRNA | AB451294.1 | - | 4 | 378 | ||
AB451428.1 | - | 4 | 378 | |||
AF124491.1 | - | 4 | 378 | |||
AK222736.1 | - | 4 | 450 | |||
AK303198.1 | - | 3 | 244 | |||
AK304240.1 | - | 4 | 527 | |||
BC001379.2 | - | 4 | 443 | 3051069 | ||
BC014223.2 | - | 3 | 294 | |||
BP871813.1 | - | 4 | 543 | |||
BT007312.1 | - | 4 | 378 | |||
CR533568.1 | - | 4 | 378 | |||
D63482.2 | - | 4 | 449 |