Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUP58 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008564.1 | NP_001008564.1 | 3 | 575 | ||
NM_014089.3 | NP_054808.1 | 4 | 611 | |||
XM_005266600.3 | XP_005266657.1 | 4 | 955 | |||
XM_011535328.1 | XP_011533630.1 | 4 | 955 | |||
XM_011535329.2 | XP_011533631.1 | 5 | 1041 | |||
XM_017020880.1 | XP_016876369.1 | 6 | 1123 | |||
XM_017020881.1 | XP_016876370.1 | 5 | 1041 | |||
GenBank mRNA | AB007870.1 | - | 4 | 508 | ||
AK123955.1 | - | 5 | 445 | |||
AK302946.1 | - | 3 | 470 | |||
AK303409.1 | - | 3 | 379 | |||
BC001104.2 | - | 4 | 483 | 3504343 | ||
BG330006.1 | - | 428 |