Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJC6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256864.1 | NP_001243793.1 | 9 | 1358 | ||
NM_001256865.1 | NP_001243794.1 | 10 | 1306 | |||
NM_014787.3 | NP_055602.1 | 9 | 1193 | |||
GenBank mRNA | AB007942.1 | - | 9 | 1133 | ||
AK296408.1 | - | 10 | 1260 | |||
BC051722.1 | - | 9 | 1142 | |||
BC109279.2 | - | 9 | 1291 | 40028812 | ||
BC109280.1 | - | 9 | 1294 | 40028814 | ||
BX647203.1 | - | 9 | 1347 |