Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PJA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014819.4 | NP_055634.3 | 4 | 1280 | ||
XM_017010099.1 | XP_016865588.1 | 5 | 1356 | |||
GenBank mRNA | AB007898.1 | - | 4 | 1177 | ||
AK291759.1 | - | 4 | 1214 | |||
BC030826.1 | - | 4 | 1176 | 4811738 | ||
BP301854.1 | - | 1 | 217 |