Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEC24D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318066.1 | NP_001304995.1 | 20 | 2907 | ||
NM_014822.3 | NP_055637.2 | 20 | 2904 | |||
XM_005263379.2 | XP_005263436.1 | 20 | 2781 | |||
XM_017008875.1 | XP_016864364.1 | 15 | 2062 | |||
GenBank mRNA | AB018298.1 | - | 20 | 2807 | ||
AF130464.2 | - | 19 | 2607 | |||
AK000709.1 | - | 1 | 514 | |||
AK001390.1 | - | 17 | 2275 | |||
AK125962.1 | - | 15 | 2739 | |||
AK291765.1 | - | 20 | 2804 | |||
AK309960.1 | - | 13 | 1643 | |||
BC035761.1 | - | 20 | 2829 | 5554774 |