Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other UBAP2L SIRNA Assays ›
Gene Symbol:
UBAP2L
Gene Name:
ubiquitin associated protein 2 like
Gene Aliases:
NICE-4, NICE4
Chromosome Location:
Chr.1: 154220172 - 154271510 on Build GRCh38
UniGene ID:
Hs.490551
Species:
Human
siRNA ID
HSS114969
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: UBAP2L
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001127320.2
NP_001120792.1
8
792
NM_001287815.1
NP_001274744.1
8
825
NM_001287816.1
NP_001274745.1
8
879
NM_014847.3
NP_055662.3
8
846
XM_005245658.3
XP_005245715.1
8
879
XM_005245667.3
XP_005245724.1
8
879
XM_005245668.3
XP_005245725.1
8
879
XM_005245669.3
XP_005245726.1
8
879
XM_005245670.3
XP_005245727.1
8
879
XM_005245672.3
XP_005245729.1
8
771
XM_005245673.3
XP_005245730.1
8
879
XM_005245674.1
XP_005245731.1
8
771
XM_011510215.2
XP_011508517.1
8
879
XM_011510216.1
XP_011508518.1
8
771
XM_017002972.1
XP_016858461.1
8
904
XM_017002973.1
XP_016858462.1
8
771
XM_017002974.1
XP_016858463.1
8
802
XM_017002975.1
XP_016858464.1
8
879
XM_017002976.1
XP_016858465.1
8
781
XM_017002977.1
XP_016858466.1
8
771
XM_017002978.1
XP_016858467.1
8
802
XM_017002979.1
XP_016858468.1
8
763
XM_017002980.1
XP_016858469.1
8
802
XM_017002981.1
XP_016858470.1
8
763
XM_017002982.1
XP_016858471.1
8
771
XM_017002983.1
XP_016858472.1
8
802
XM_017002984.1
XP_016858473.1
8
802
XM_017002985.1
XP_016858474.1
8
904
XM_017002986.1
XP_016858475.1
8
771
XM_017002987.1
XP_016858476.1
8
802
XM_017002988.1
XP_016858477.1
8
802
XM_017002989.1
XP_016858478.1
8
802
XM_017002990.1
XP_016858479.1
8
763
XM_017002991.1
XP_016858480.1
7
704
GenBank mRNA
AK302662.1
-
8
813
AK302953.1
-
5
540
AK303533.1
-
8
752
BC003170.1
-
7
728
2906083
D63478.1
-
8
785
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
binding of sperm to zona pellucida
hematopoietic stem cell homeostasis
protein binding
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-jmrqp:80/10.239.165.107:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0