Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IQSEC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134382.2 | NP_001127854.1 | 11 | 2663 | ||
NM_014869.6 | NP_055684.3 | 11 | 2849 | |||
XM_011534304.2 | XP_011532606.1 | 13 | 2900 | |||
XM_011534305.2 | XP_011532607.1 | 11 | 2649 | |||
XM_011534306.2 | XP_011532608.1 | 11 | 3055 | |||
XM_011534307.2 | XP_011532609.1 | 12 | 3001 | |||
XM_011534308.2 | XP_011532610.1 | 11 | 2844 | |||
XM_011534310.2 | XP_011532612.1 | 13 | 2900 | |||
XM_011534311.2 | XP_011532613.1 | 11 | 3012 | |||
XM_011534312.2 | XP_011532614.1 | 11 | 2640 | |||
XM_011534313.2 | XP_011532615.1 | 11 | 2859 | |||
XM_011534314.2 | XP_011532616.1 | 11 | 2595 | |||
XM_011534315.2 | XP_011532617.1 | 11 | 2598 | |||
XM_017007581.1 | XP_016863070.1 | 12 | 2870 | |||
XM_017007582.1 | XP_016863071.1 | 12 | 2855 | |||
GenBank mRNA | AB018306.1 | - | 10 | 2342 | ||
AK294525.1 | - | 11 | 2663 | |||
AY653734.1 | - | 11 | 2611 | |||
BC010267.1 | - | 11 | 2603 | 2900367 |