Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HELZ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014877.3 | NP_055692.2 | 19 | 2588 | ||
XM_005257888.4 | XP_005257945.1 | 20 | 2717 | |||
XM_005257889.4 | XP_005257946.1 | 19 | 2639 | |||
XM_006722214.3 | XP_006722277.1 | 20 | 2720 | |||
XM_006722215.3 | XP_006722278.1 | 18 | 2592 | |||
XM_006722216.3 | XP_006722279.1 | 9 | 1422 | |||
XM_011525544.2 | XP_011523846.1 | 20 | 2791 | |||
XM_017025477.1 | XP_016880966.1 | 17 | 2508 | |||
XM_017025478.1 | XP_016880967.1 | 20 | 2773 | |||
GenBank mRNA | BC094881.1 | - | 17 | 2458 | 3076113 | |
BC130582.1 | - | 19 | 2568 | 40146613 | ||
BC144083.1 | - | 19 | 2571 | 9052603 | ||
D29677.2 | - | 18 | 2546 |