Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGAP25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007231.2 | NP_001007232.2 | 11 | 2309 | ||
NM_001166276.1 | NP_001159748.1 | 10 | 2249 | |||
NM_001166277.1 | NP_001159749.1 | 10 | 2189 | |||
NM_014882.2 | NP_055697.1 | 10 | 2246 | |||
XM_005264675.2 | XP_005264732.1 | 11 | 2013 | |||
XM_005264676.2 | XP_005264733.1 | 10 | 1897 | |||
XM_011533206.2 | XP_011531508.1 | 11 | 2057 | |||
XM_011533207.2 | XP_011531509.1 | 11 | 2023 | |||
XM_011533208.2 | XP_011531510.1 | 12 | 2074 | |||
XM_011533209.2 | XP_011531511.1 | 8 | 1614 | |||
XM_011533210.2 | XP_011531512.1 | 6 | 1141 | |||
XM_017005423.1 | XP_016860912.1 | 11 | 1917 | |||
XM_017005424.1 | XP_016860913.1 | 11 | 1914 | |||
XM_017005425.1 | XP_016860914.1 | 10 | 1800 | |||
XM_017005426.1 | XP_016860915.1 | 8 | 2251 | |||
XM_017005427.1 | XP_016860916.1 | 8 | 2248 | |||
GenBank mRNA | AK290396.1 | - | 11 | 1987 | ||
AK297056.1 | - | 10 | 2067 | |||
AK297157.1 | - | 11 | 2049 | |||
D29642.1 | - | 10 | 2059 | |||
EU794656.1 | - | 10 | 2246 |